Product Details

SNP ID
rs37024
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:56932159 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGGCGCGCGCCCCAGAGACGTGAAC[G/T]GTCGTTGCAGAGATTGCGGGCGGCT
Phenotype
MIM: 608070
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HERPUD1 PubMed Links

Gene Details

Gene
HERPUD1
Gene Name
homocysteine inducible ER protein with ubiquitin like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010989.2 70 UTR 5 NP_001010989.1
NM_001272103.1 70 UTR 5 NP_001259032.1
NM_014685.3 70 UTR 5 NP_055500.1

View Full Product Details