Product Details

SNP ID
rs30020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23393034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGGAAATACCAGTGCGTTCC[A/G]CAGTGGGATGTGGTTATAGATTTTT
Phenotype
MIM: 606978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG7 PubMed Links

Gene Details

Gene
COG7
Gene Name
component of oligomeric golgi complex 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153603.3 Intron NP_705831.1
XM_017023870.1 Intron XP_016879359.1

View Full Product Details