Product Details

SNP ID
rs2362482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5949747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGTATCCGAACACGGAGGCTCA[A/G]CCCTCCACACCCAGCCCTTATTGAA
Phenotype
MIM: 610109 MIM: 613160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO2 PubMed Links

Gene Details

Gene
ANO2
Gene Name
anoctamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278596.1 Intron NP_001265525.1
NM_001278597.1 Intron NP_001265526.1
XM_006718988.3 Intron XP_006719051.1
XM_011520975.2 Intron XP_011519277.1
XM_011520978.2 Intron XP_011519280.1
XM_017019672.1 Intron XP_016875161.1
Gene
VWF
Gene Name
von Willebrand factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000552.4 Intron NP_000543.2

View Full Product Details