Product Details

SNP ID
rs61751305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5949823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATCCACCTCTACTTCAGACTTA[C/T]AGCTTCCCACCTTGACATACTGCAG
Phenotype
MIM: 610109 MIM: 613160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO2 PubMed Links

Gene Details

Gene
ANO2
Gene Name
anoctamin 2
There are no transcripts associated with this gene.

Gene
VWF
Gene Name
von Willebrand factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000552.4 8471 Missense Mutation TAT,TGT Y2739C NP_000543.2

View Full Product Details