Product Details

SNP ID
rs121907981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72345550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCGGCAACAGCCCCTGCTCTGGG[C/G]CTGGAGGAAAAGGGGCATGTGCCAG
Phenotype
MIM: 606869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HEXA PubMed Links

Gene Details

Gene
HEXA
Gene Name
hexosaminidase subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000520.5 1956 Missense Mutation TGC,TGG C474W NP_000511.2
NM_001318825.1 1956 Missense Mutation TGC,TGG C485W NP_001305754.1

View Full Product Details