Product Details

SNP ID
rs11760176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30002650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCATGTCCAATCTCTTAGATTCA[C/T]TCAATGGGGTATTTCGCATATTTTA
Phenotype
MIM: 615801 MIM: 615714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCG8 PubMed Links

Gene Details

Gene
HCG8
Gene Name
HLA complex group 8
There are no transcripts associated with this gene.

Gene
HLA-J
Gene Name
major histocompatibility complex, class I, J (pseudogene)
There are no transcripts associated with this gene.

Gene
ZNRD1ASP
Gene Name
zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.

View Full Product Details