Product Details

SNP ID
rs11753939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30071990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAGAGGTTGTCTCGTTTGTGG[C/G]GTTGGGGGAGGAAGAGTGAGGCTGA
Phenotype
MIM: 606670 MIM: 607524 MIM: 607525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R11 PubMed Links

Gene Details

Gene
PPP1R11
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 11
There are no transcripts associated with this gene.

Gene
RNF39
Gene Name
ring finger protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025236.3 Intron NP_079512.2
NM_170769.2 Intron NP_739575.2
XM_017011325.1 Intron XP_016866814.1
XM_017011326.1 Intron XP_016866815.1
Gene
ZNRD1
Gene Name
zinc ribbon domain containing 1
There are no transcripts associated with this gene.

View Full Product Details