Product Details

SNP ID
rs7153068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34990275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAAGAGGAATATACATCTTTAA[A/T]TATGATTAAAATGTGATTTCTTCTA
Phenotype
MIM: 604857
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SRP54 PubMed Links

Gene Details

Gene
SRP54
Gene Name
signal recognition particle 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146282.1 Intron NP_001139754.1
NM_003136.3 Intron NP_003127.1
XM_005268024.2 Intron XP_005268081.1
XM_011537106.1 Intron XP_011535408.1
XM_017021615.1 Intron XP_016877104.1

View Full Product Details