Product Details
- SNP ID
-
rs12483924
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36800861 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGATTGGGTGTTCAGGGCAGAGA[C/G]GTGGAAGACCAGGGGCAGTCAGTGC
- Phenotype
-
MIM: 168890
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PVALB
PubMed Links
Gene Details
- Gene
- PVALB
- Gene Name
- parvalbumin
View Full Product Details