Product Details

SNP ID
rs12484441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37701352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGATCATAGGAACTGCCCTGGC[C/G]TGACTCACCCAATATGGAGGAGGTG
Phenotype
MIM: 609761
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOL12 PubMed Links

Gene Details

Gene
NOL12
Gene Name
nucleolar protein 12
There are no transcripts associated with this gene.

Gene
TRIOBP
Gene Name
TRIO and F-actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039141.2 258 UTR 5 NP_001034230.1
NM_007032.5 258 Intron NP_008963.3
NM_138632.2 258 Intron NP_619538.2

View Full Product Details