Product Details
- SNP ID
-
rs2072081
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44250125 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACCCCACCCGCTCACCCACCTCCT[G/T]CCTTTGCTTCTACCCCTGCCTGTGC
- Phenotype
-
MIM: 109270
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC4A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13306777] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC4A1
- Gene Name
- solute carrier family 4 member 1 (Diego blood group)
View Full Product Details