Product Details

SNP ID
rs10416809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1252312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCTGCTGAGGGCTGGGGGTGG[C/T]GGCTTGGGTGACACTCCTTCCGCCT
Phenotype
MIM: 603150 MIM: 606700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5D PubMed Links

Gene Details

Gene
ATP5D
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
There are no transcripts associated with this gene.

Gene
MIDN
Gene Name
midnolin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177401.4 Intron NP_796375.3
XM_005259671.3 Intron XP_005259728.1
XM_005259672.3 Intron XP_005259729.1

View Full Product Details