Product Details
- SNP ID
-
rs758145
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2073467 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GACATCAGCTCCCACCAGCCACTCC[C/T]AAAAGGGGTGATGCGAACAGCAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MOB3A
PubMed Links
Gene Details
- Gene
- MOB3A
- Gene Name
- MOB kinase activator 3A
View Full Product Details