Product Details

SNP ID
rs502121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1387300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCCAGGGAGGCCCCAGCATTGC[C/T]GGAGTGGCAGGCACCCCACAAGCAC
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
There are no transcripts associated with this gene.

Gene
MUM1
Gene Name
melanoma associated antigen (mutated) 1
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 Intron NP_077718.3
XM_005259556.4 Intron XP_005259613.2
XM_017026768.1 Intron XP_016882257.1

View Full Product Details