Product Details

SNP ID
rs34395206
Assay Type
Functionally Tested
NCBI dbSNP Submissions
31
Location
Chr.1:155267848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTAAGGAATGTCATGGGCCCCC[C/T]CCCTTTCCTACCCTGATGTTCTACT
Phenotype
MIM: 602989 MIM: 609973 MIM: 606913
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links
Additional Information
For this assay, SNP(s) [rs150618341] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294338.1 Intron NP_001281267.1
NM_001294339.1 Intron NP_001281268.1
NM_003993.3 Intron NP_003984.2
XM_005244878.1 Intron XP_005244935.1
XM_011509143.1 Intron XP_011507445.1
XM_011509144.1 Intron XP_011507446.1
XM_011509147.2 Intron XP_011507449.1
XM_017000241.1 Intron XP_016855730.1
XM_017000242.1 Intron XP_016855731.1
XM_017000243.1 Intron XP_016855732.1
XM_017000244.1 Intron XP_016855733.1
Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
There are no transcripts associated with this gene.

Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
There are no transcripts associated with this gene.

View Full Product Details