Product Details

SNP ID
rs2191212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68976109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATACATGTATATTCCATGGATG[A/T]TATAGAATCACTAAAAATGACCCAC
Phenotype
MIM: 612507
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCA9 PubMed Links

Gene Details

Gene
ABCA9
Gene Name
ATP binding cassette subfamily A member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080283.3 Intron NP_525022.2
XM_017024011.1 Intron XP_016879500.1
XM_017024012.1 Intron XP_016879501.1
XM_017024013.1 Intron XP_016879502.1
XM_017024014.1 Intron XP_016879503.1
XM_017024015.1 Intron XP_016879504.1

View Full Product Details