Product Details

SNP ID
rs6956431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92120702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGATGACCTCACATCATATACA[C/T]AAATTAATTCATAATGGATCAAAGA
Phenotype
MIM: 601637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP51A1 PubMed Links

Gene Details

Gene
CYP51A1
Gene Name
cytochrome P450 family 51 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000786.3 Intron NP_000777.1
NM_001146152.1 Intron NP_001139624.1

View Full Product Details