Product Details

SNP ID
rs10216002
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:77699012 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGATTCTTAAATAGCGGTATTAACA[A/C]GTGGTATAGCAAGAATACATATATG
Phenotype
MIM: 616048
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
APTR PubMed Links
Additional Information
For this assay, SNP(s) [rs117613808] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
APTR
Gene Name
Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
RSBN1L
Gene Name
round spermatid basic protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198467.2 Intron NP_940869.2

View Full Product Details