Product Details

SNP ID
rs10459545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73952506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGGACTGCAGTTGCACAAACAT[A/G]GCTCAGGTGTGCCACCATGCCTGGC
Phenotype
MIM: 614647 MIM: 603162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ6 PubMed Links

Gene Details

Gene
COQ6
Gene Name
coenzyme Q6, monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182476.2 Intron NP_872282.1
NM_182480.2 Intron NP_872286.2
XM_006720156.1 Intron XP_006720219.1
XM_011536807.1 Intron XP_011535109.1
XM_011536808.1 Intron XP_011535110.1
XM_011536809.2 Intron XP_011535111.1
XM_011536810.2 Intron XP_011535112.1
XM_017021351.1 Intron XP_016876840.1
XM_017021352.1 Intron XP_016876841.1
Gene
ENTPD5
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
There are no transcripts associated with this gene.

Gene
FAM161B
Gene Name
family with sequence similarity 161 member B
There are no transcripts associated with this gene.

View Full Product Details