Product Details
- SNP ID
-
rs11976307
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:38248573 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGAAAATGGAAGAATGTTTGTACA[A/T]GTAAGCCTTACCAACCTGTAATTAT
- Phenotype
-
MIM: 609642
MIM: 615450
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TARP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78514022] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TARP
- Gene Name
- TCR gamma alternate reading frame protein
- Gene
- TRGC2
- Gene Name
- T cell receptor gamma constant 2
There are no transcripts associated with this gene.
View Full Product Details