Product Details

SNP ID
rs2014626
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2714773 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCCAGCAGCCAGGACAGGCTCGGG[C/T]CCCCTCCTATTGCAGATGCCCTCTC
Phenotype
MIM: 607862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRAS1 PubMed Links

Gene Details

Gene
DIRAS1
Gene Name
DIRAS family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145173.3 3357 UTR 3 NP_660156.1
XM_011527713.2 3357 UTR 3 XP_011526015.1

View Full Product Details