Product Details

SNP ID
rs914172
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:41142741 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGATCTGACCAACCCTCCTCCTGAA[C/G]AGGAGTTGGAAGATGGTCTCCAGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00323 PubMed Links

Gene Details

Gene
LINC00323
Gene Name
long intergenic non-protein coding RNA 323
There are no transcripts associated with this gene.

View Full Product Details