Product Details

SNP ID
rs953741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178902845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTTGATAACAGATAATATTTAC[C/T]CAGGAGTCTGCACCTGTTCTATTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107984119 PubMed Links

Gene Details

Gene
LOC107984119
Gene Name
zinc finger protein 181-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017010140.1 Intron XP_016865629.1
Gene
ZFP2
Gene Name
ZFP2 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030613.3 Intron NP_085116.2
Gene
ZNF354B
Gene Name
zinc finger protein 354B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058230.2 Intron NP_478137.1

View Full Product Details