Product Details

SNP ID
rs2097020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:66501779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATGTGAAATAGGTTGATTGGAC[A/G]TCATTTTACAAAGTGAGATCTGGAA
Phenotype
MIM: 603016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH19 PubMed Links

Gene Details

Gene
CDH19
Gene Name
cadherin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271028.1 5538 UTR 3 NP_001257957.1
NM_021153.3 5538 UTR 3 NP_066976.1
XM_011525931.2 5538 UTR 3 XP_011524233.1
XM_011525932.1 5538 Intron XP_011524234.1
XM_017025711.1 5538 UTR 3 XP_016881200.1

View Full Product Details