Product Details

SNP ID
rs8182245
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6859252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTGTTGAAAATTTTTGGATCTG[C/T]GTTCATCAAGGATGTTGGTCTGAAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALOX12P2 PubMed Links

Gene Details

Gene
ALOX12P2
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details