Product Details

SNP ID
rs1019398
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:4435898 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGGATTTCCTGCTCTTGTGGGGAA[A/G]CAGCTGGGTCGGGGCTAGAAGAACA
Phenotype
MIM: 134921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF6 PubMed Links

Gene Details

Gene
FGF6
Gene Name
fibroblast growth factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020996.2 Intron NP_066276.2
XM_017018995.1 Intron XP_016874484.1
XM_017018996.1 Intron XP_016874485.1

View Full Product Details