Product Details
- SNP ID
-
rs7284093
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50683093 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CACACAGGGGCTTCACATGTGTGCA[T/C]GTGTGTTCTGAATAACCAGGTATGA
- Phenotype
-
MIM: 606230
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SHANK3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73429494,rs9616933] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHANK3
- Gene Name
- SH3 and multiple ankyrin repeat domains 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033517.1 |
|
Intron |
|
|
NP_277052.1 |
View Full Product Details