Product Details

SNP ID
rs4784881
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58007174 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGAGAGATATGGGTCTGTAGTTT[C/T]CTTGTAATGTCTTTTTCTGGTATTG
Phenotype
MIM: 613276
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
USB1 PubMed Links

Gene Details

Gene
USB1
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195302.1 Intron NP_001182231.1
NM_001204911.1 Intron NP_001191840.1
NM_024598.3 Intron NP_078874.2
XM_005256144.3 Intron XP_005256201.1
XM_011523328.2 Intron XP_011521630.1
XM_011523329.2 Intron XP_011521631.1
XM_011523330.1 Intron XP_011521632.1
Gene
ZNF319
Gene Name
zinc finger protein 319
There are no transcripts associated with this gene.

View Full Product Details