Product Details

SNP ID
rs12445769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56932874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTGGCTCTTCTCCTCCCTGGCC[A/G]CCTTCCGCCGTGCCCCTCCCTGTCG
Phenotype
MIM: 608070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HERPUD1 PubMed Links

Gene Details

Gene
HERPUD1
Gene Name
homocysteine inducible ER protein with ubiquitin like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010989.2 Intron NP_001010989.1
NM_001272103.1 Intron NP_001259032.1
NM_014685.3 Intron NP_055500.1

View Full Product Details