Product Details
- SNP ID
-
rs6559872
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:85550131 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCAGGCTGAGTGTGCAAAGAGGAG[G/T]AAGTTAGCGGTGCAAAGAGGTATAC
- Phenotype
-
MIM: 606830
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGTPBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35611324] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGTPBP1
- Gene Name
- ATP/GTP binding protein 1
View Full Product Details