Product Details

SNP ID
rs7071652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94002839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGATCACATGATAAATACATCATA[C/T]GCAATTTTGGATCCTAGATTCCTTG
Phenotype
MIM: 608414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCE1 PubMed Links

Gene Details

Gene
PLCE1
Gene Name
phospholipase C epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165979.2 Intron NP_001159451.1
NM_001288989.1 Intron NP_001275918.1
NM_016341.3 Intron NP_057425.3
XM_006717885.3 Intron XP_006717948.1
XM_006717888.3 Intron XP_006717951.1
XM_006717889.3 Intron XP_006717952.1
XM_006717890.2 Intron XP_006717953.1
XM_011539849.2 Intron XP_011538151.1
XM_011539850.2 Intron XP_011538152.1
XM_011539851.2 Intron XP_011538153.1
XM_011539852.2 Intron XP_011538154.1
XM_017016310.1 Intron XP_016871799.1
XM_017016311.1 Intron XP_016871800.1
XM_017016312.1 Intron XP_016871801.1

View Full Product Details