Product Details
- SNP ID
-
hCV2985358
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46091918 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTGGGCCAGCCCCTCCCCGGCT[A/C]TGAGCCTCAGCTTCCCACAGTCTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3619
PubMed Links
Gene Details
- Gene
- MIR3619
- Gene Name
- microRNA 3619
There are no transcripts associated with this gene.
- Gene
- MIRLET7BHG
- Gene Name
- MIRLET7B host gene
There are no transcripts associated with this gene.
View Full Product Details