Product Details

SNP ID
rs574462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122404863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAATAAGCAGAGATAGAGTGAT[A/G]TGAATGAATAAGGAGTGTATGTGAG
Phenotype
MIM: 140581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSF2 PubMed Links

Gene Details

Gene
HSF2
Gene Name
heat shock transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135564.1 Intron NP_001129036.1
NM_001243094.1 Intron NP_001230023.1
NM_004506.3 Intron NP_004497.1

View Full Product Details