Product Details

SNP ID
rs4672926
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:219373774 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAGTGGATGATGAAGGGTGATAAG[A/G]TATAGACTGTGCCCTTCTCTTTGCA
Phenotype
MIM: 611367
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNPEP PubMed Links

Gene Details

Gene
DNPEP
Gene Name
aspartyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319116.1 2014 UTR 3 NP_001306045.1
NM_001319117.1 2014 UTR 3 NP_001306046.1
NM_001319118.1 2014 UTR 3 NP_001306047.1
NM_001319119.1 2014 UTR 3 NP_001306048.1
NM_001319120.1 2014 UTR 3 NP_001306049.1
NM_001319121.1 2014 Intron NP_001306050.1
NM_001319122.1 2014 UTR 3 NP_001306051.1
NM_012100.3 2014 UTR 3 NP_036232.2

View Full Product Details