Product Details

SNP ID
rs12522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5290283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAAGCAACCATTATTTGTCCACA[A/G]AAACAGTGAAAAACAGTGATATAAA
Phenotype
MIM: 602126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB14 PubMed Links

Gene Details

Gene
ZBTB14
Gene Name
zinc finger and BTB domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143823.2 2087 UTR 3 NP_001137295.1
NM_001243702.1 2087 UTR 3 NP_001230631.1
NM_001243704.1 2087 UTR 3 NP_001230633.1
NM_003409.4 2087 UTR 3 NP_003400.2

View Full Product Details