Product Details
- SNP ID
-
rs12930464
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:22298554 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGGTCACAGATGCCAAGCAGCTG[C/T]GGTGGGATTTGAACCTGCACAGTTT
- Phenotype
-
MIM: 606968
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EEF2K
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80036167] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EEF2K
- Gene Name
- eukaryotic elongation factor 2 kinase
There are no transcripts associated with this gene.
- Gene
- POLR3E
- Gene Name
- RNA polymerase III subunit E
View Full Product Details