Product Details

SNP ID
rs9475807
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:13934366 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCGTGCCCACCAGTGTTTGAGATA[C/T]CTCCTTTCATTCTGTTGGATTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF182 PubMed Links

Gene Details

Gene
RNF182
Gene Name
ring finger protein 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165032.1 Intron NP_001158504.1
NM_001165033.1 Intron NP_001158505.1
NM_001165034.1 Intron NP_001158506.1
NM_152737.3 Intron NP_689950.1
XM_017010448.1 Intron XP_016865937.1
XM_017010449.1 Intron XP_016865938.1
XM_017010450.1 Intron XP_016865939.1

View Full Product Details