Product Details

SNP ID
rs1747683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13319976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTACACTGAAACAAATATTGACA[A/G]ACAAAAGGGAACTTTTGATTCTCTA
Phenotype
MIM: 600902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPHS1 PubMed Links

Gene Details

Gene
SEPHS1
Gene Name
selenophosphate synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195602.1 Intron NP_001182531.1
NM_001195604.1 Intron NP_001182533.1
NM_012247.4 Intron NP_036379.2
XM_017015942.1 Intron XP_016871431.1
XM_017015943.1 Intron XP_016871432.1
XM_017015944.1 Intron XP_016871433.1
XM_017015945.1 Intron XP_016871434.1

View Full Product Details