Product Details

SNP ID
rs615530
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:37696022 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCCAGTCCCCAGATCCAACTAATAG[A/G]TGCTTACTGTGGAAGAGGTTTCATA
Phenotype
MIM: 614799 MIM: 609977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf109 PubMed Links

Gene Details

Gene
C1orf109
Gene Name
chromosome 1 open reading frame 109
There are no transcripts associated with this gene.

Gene
CDCA8
Gene Name
cell division cycle associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256875.1 Intron NP_001243804.1
NM_018101.3 Intron NP_060571.1

View Full Product Details