Product Details

SNP ID
rs34351561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100360175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTAATCAGCTAAAGGTAAAATA[A/G]TATTTTAGAAGTGCAGGAAGAAAGA
Phenotype
MIM: 607557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC17A8 PubMed Links

Gene Details

Gene
SLC17A8
Gene Name
solute carrier family 17 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145288.1 Intron NP_001138760.1
NM_139319.2 Intron NP_647480.1

View Full Product Details