Product Details

SNP ID
rs9397799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:155274564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCTTCATTTGGGCCAGGAAAT[G/T]ATTCTGGTCCCAATTAGAAAGGACC
Phenotype
MIM: 607033
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN20 PubMed Links

Gene Details

Gene
CLDN20
Gene Name
claudin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001346.3 Intron NP_001001346.1
XM_011535848.2 Intron XP_011534150.1
Gene
TFB1M
Gene Name
transcription factor B1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016020.3 Intron NP_057104.2
XM_005267006.4 Intron XP_005267063.1
XM_011535870.1 Intron XP_011534172.1
XM_011535871.1 Intron XP_011534173.1
XM_011535872.2 Intron XP_011534174.1
XM_011535873.2 Intron XP_011534175.1
XM_017010917.1 Intron XP_016866406.1

View Full Product Details