Product Details
- SNP ID
-
rs1041098
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:155264997 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATTGTTGTATAAAGGTTTTCTGTTA[C/T]ATTTAATCTGGAGTTCCTGGATGAT
- Phenotype
-
MIM: 607033
MIM: 604709
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12524284] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLDN20
- Gene Name
- claudin 20
- Gene
- TFB1M
- Gene Name
- transcription factor B1, mitochondrial
- Gene
- TIAM2
- Gene Name
- T-cell lymphoma invasion and metastasis 2
There are no transcripts associated with this gene.
View Full Product Details