Product Details

SNP ID
rs7546838
Assay Type
Validated
NCBI dbSNP Submissions
27
Location
Chr.1:156815979 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGGGAGAGATGAGGGAGCTGCACC[A/G]CGCTGCCGCCCCAGGTTTCCACTCA
Phenotype
MIM: 191315 MIM: 604514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTRK1 PubMed Links

Gene Details

Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 Intron NP_001007793.1
NM_001012331.1 Intron NP_001012331.1
NM_002529.3 Intron NP_002520.2
Gene
SH2D2A
Gene Name
SH2 domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161441.1 Intron NP_001154913.1
NM_001161442.1 Intron NP_001154914.1
NM_001161443.1 Intron NP_001154915.1
NM_001161444.1 Intron NP_001154916.1
NM_003975.3 Intron NP_003966.2
XM_006711615.2 Intron XP_006711678.1
XM_017002762.1 Intron XP_016858251.1
XM_017002763.1 Intron XP_016858252.1
XM_017002764.1 Intron XP_016858253.1
XM_017002765.1 Intron XP_016858254.1
XM_017002766.1 Intron XP_016858255.1
XM_017002767.1 Intron XP_016858256.1

View Full Product Details