Product Details

SNP ID
rs2036215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:27725661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATCAGGGAGGTTCTGAAGCTT[C/T]TTTAAAGGCCCTTACTTTTCACTTG
Phenotype
MIM: 611409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCA2 PubMed Links

Gene Details

Gene
OCA2
Gene Name
OCA2 melanosomal transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000275.2 Intron NP_000266.2
NM_001300984.1 Intron NP_001287913.1
XM_011521640.2 Intron XP_011519942.1
XM_017022255.1 Intron XP_016877744.1
XM_017022256.1 Intron XP_016877745.1
XM_017022257.1 Intron XP_016877746.1
XM_017022258.1 Intron XP_016877747.1
XM_017022259.1 Intron XP_016877748.1
XM_017022260.1 Intron XP_016877749.1
XM_017022261.1 Intron XP_016877750.1
XM_017022262.1 Intron XP_016877751.1
XM_017022263.1 Intron XP_016877752.1
XM_017022264.1 Intron XP_016877753.1
XM_017022265.1 Intron XP_016877754.1

View Full Product Details