Product Details

SNP ID
rs3808317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:157151766 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGGGCTATGCAATTGGACCAAG[A/G]GGAGTCAATGATTTTGAAGTCAGTC
Phenotype
MIM: 614454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE3C PubMed Links

Gene Details

Gene
UBE3C
Gene Name
ubiquitin protein ligase E3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014671.2 Intron NP_055486.2
XM_005249564.4 Intron XP_005249621.1
XM_017012818.1 Intron XP_016868307.1

View Full Product Details