Product Details

SNP ID
rs34315859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41404336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGTGTCACCAAGTACTGTATTA[A/C]CTATGATTGCTGGAATGAACTGGAT
Phenotype
MIM: 616114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHD6 PubMed Links

Gene Details

Gene
CHD6
Gene Name
chromodomain helicase DNA binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032221.4 8038 UTR 3 NP_115597.3
XM_005260573.2 8038 UTR 3 XP_005260630.1
XM_005260576.4 8038 UTR 3 XP_005260633.1
XM_011529080.2 8038 UTR 3 XP_011527382.1
XM_011529082.2 8038 UTR 3 XP_011527384.1
XM_017028099.1 8038 UTR 3 XP_016883588.1
XM_017028100.1 8038 UTR 3 XP_016883589.1
XM_017028101.1 8038 UTR 3 XP_016883590.1
XM_017028102.1 8038 Intron XP_016883591.1
XM_017028103.1 8038 UTR 3 XP_016883592.1
XM_017028104.1 8038 Intron XP_016883593.1

View Full Product Details