Product Details

SNP ID
rs3743851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1974130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTACCAGCCTGGGGAAGGGCAG[C/T]GGGGCGGGCAGCCAGAGGCCGCGGG
Phenotype
MIM: 611256 MIM: 603624 MIM: 605915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOXO1 PubMed Links

Gene Details

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
There are no transcripts associated with this gene.

Gene
RNF151
Gene Name
ring finger protein 151
There are no transcripts associated with this gene.

Gene
RPS2
Gene Name
ribosomal protein S2
There are no transcripts associated with this gene.

Gene
SNHG9
Gene Name
small nucleolar RNA host gene 9
There are no transcripts associated with this gene.

Gene
SNORA78
Gene Name
small nucleolar RNA, H/ACA box 78
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006453.2 Intron NP_006444.2

View Full Product Details