Product Details

SNP ID
rs3827416
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:152833907 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTGATAACTTGTGAATGAGGAAAT[T/C]AATGCATGAGGTGACACCTGGGGGT
Phenotype
MIM: 300006 MIM: 300275
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CETN2 PubMed Links

Gene Details

Gene
CETN2
Gene Name
centrin 2
There are no transcripts associated with this gene.

Gene
NSDHL
Gene Name
NAD(P) dependent steroid dehydrogenase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129765.1 Intron NP_001123237.1
NM_015922.2 Intron NP_057006.1
XM_011531178.2 Intron XP_011529480.1
XM_017029564.1 Intron XP_016885053.1

View Full Product Details