Product Details

SNP ID
rs708220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31384756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATAGGCCGCAAATCCAACTTTG[C/G]CTGCAGTTTTTTTCTTTTTTTTTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DENND5B PubMed Links

Gene Details

Gene
DENND5B
Gene Name
DENN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308339.1 6971 UTR 3 NP_001295268.1
NM_144973.3 6971 UTR 3 NP_659410.3
XM_005253326.2 6971 UTR 3 XP_005253383.1
XM_005253327.3 6971 UTR 3 XP_005253384.1
XM_005253330.3 6971 UTR 3 XP_005253387.1
XM_006719040.3 6971 UTR 3 XP_006719103.1
XM_011520590.2 6971 UTR 3 XP_011518892.1
XM_017018896.1 6971 UTR 3 XP_016874385.1
XM_017018897.1 6971 UTR 3 XP_016874386.1
XM_017018898.1 6971 UTR 3 XP_016874387.1
XM_017018899.1 6971 UTR 3 XP_016874388.1

View Full Product Details