Product Details

SNP ID
rs975496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35265666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTCAAGCATACATCTAGGGGAA[A/G]TTTATTCCAGTCAGGAAGAACAGCA
Phenotype
MIM: 194549
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF76 PubMed Links

Gene Details

Gene
ZNF76
Gene Name
zinc finger protein 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292032.1 Intron NP_001278961.1
NM_003427.4 Intron NP_003418.2
XM_011514846.2 Intron XP_011513148.1
XM_011514847.2 Intron XP_011513149.1
XM_011514849.2 Intron XP_011513151.1
XM_011514850.2 Intron XP_011513152.1
XM_011514854.2 Intron XP_011513156.1
XM_011514855.2 Intron XP_011513157.1
XM_011514856.2 Intron XP_011513158.1
XM_017011248.1 Intron XP_016866737.1
XM_017011249.1 Intron XP_016866738.1
XM_017011250.1 Intron XP_016866739.1
XM_017011251.1 Intron XP_016866740.1
XM_017011252.1 Intron XP_016866741.1
XM_017011253.1 Intron XP_016866742.1
XM_017011254.1 Intron XP_016866743.1
XM_017011255.1 Intron XP_016866744.1
XM_017011256.1 Intron XP_016866745.1
XM_017011257.1 Intron XP_016866746.1

View Full Product Details